Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG2b Kappa Light Chain, Clone: 149/10H5, Biotin, Isotype control, Novus Biologicals™
SDP

Catalog No. NBP1435635 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.1 mg
0.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1435635 0.5 mg
NBP14356305 0.05 mg
NBP14356301 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP1435635 Supplier Novus Biologicals Supplier No. NBP1435630.5MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IgG2b Kappa Light Chain Monoclonal specifically detects IgG2b Kappa Light Chain in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG2b Kappa Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone 149/10H5
Conjugate Biotin
Dilution Flow Cytometry 1:10 - 1:1000
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide
Host Species Rat
Quantity 0.5 mg
Primary or Secondary Primary
Test Specificity The monoclonal rat IgG2b, kappa immunoglobulin is useful as an isotype control for rat IgG2b antibodies.
Target Species Rat
Content And Storage Store at 4C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.