Learn More
Rat IgG2b Kappa Light Chain Isotype Control (149/10H5), Alexa Fluor™ 488, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | IgG2b Kappa Light Chain |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 149/10H5 |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry 1:10 - 1:1000 |
| Formulation | 50 mM Sodium Borate. with 0.05% Sodium Azide |
| Host Species | Rat |
| Purification Method | Protein A or G purified |
| Quantity | 500 Tests |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.