Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG2b Kappa Light Chain Isotype Control (149/10H5), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP143323V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143323V 500 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143323V Supplier Novus Biologicals Supplier No. NBP143323V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IgG2b Kappa Light Chain Monoclonal specifically detects IgG2b Kappa Light Chain in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen IgG2b Kappa Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone 149/10H5
Conjugate DyLight 405
Dilution Flow Cytometry 1:10 - 1:1000
Formulation 50 mM Sodium Borate. with 0.05% Sodium Azide
Host Species Rat
Quantity 500 Tests
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.