Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IL-1 beta/IL-1F2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF501SP
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Polyclonal specifically detects IL-1 beta/IL-1F2 in Rat samples. It is validated for Western Blot, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).Specifications
IL-1 beta/IL-1F2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Goat | |
Affinity Purified | |
RUO | |
3553 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization, Immunocytochemistry, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.5-2.0 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Q63264 | |
IL1B | |
E. coli-derived recombinant rat IL-1 beta/IL-1F2 (R&D Systems, Catalog # 501-RL) Val117-Ser268 Accession # Q63264 | |
25 μg | |
Primary | |
Detects rat IL-1 beta/IL-1F2 in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant mouse IL-1 beta is observed and less than 0.5% cross-reactivity with recombinant human IL-1 beta and recombinant porcine IL-1 beta is observed. | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction