Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IL-1 beta/IL-1F2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
$200.50 - $2020.00
Specifications
Antigen | IL-1 beta/IL-1F2 |
---|---|
Clone | 38123 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB5011SP
![]() |
R&D Systems
MAB5011SP |
25 μg |
Each for $200.50
|
|
|||||
MAB5011100
![]() |
R&D Systems
MAB5011100 |
100 μg |
Each for $549.50
|
|
|||||
MAB5011500
![]() |
R&D Systems
MAB5011500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Rat samples. It is validated for Western Blot.Specifications
IL-1 beta/IL-1F2 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Mouse | |
Rat | |
Q63264 | |
3553 | |
E. coli-derived recombinant rat IL-1 beta Val117-Ser268 Accession # Q63264 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat IL-1 beta in direct ELISAs and Western blots. In these formats, this antibody shows approximately 100% cross-reactivity with recombinant mouse (rm) IL-1 beta and rcrIL-1 beta and no cross-reactivity with rfIL-1 beta , rhIL-1 beta , or rrmIL-1 beta . |
38123 | |
Western Blot | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
IL1B | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title