Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IL-1 beta/IL-1F2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF501
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF501 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF501 Supplier R&D Systems Supplier No. BAF501
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

IL-1 beta/IL-1F2 Polyclonal specifically detects IL-1 beta/IL-1F2 in Rat samples. It is validated for Western Blot, Immunocytochemistry, ELISA Detection (Matched Antibody Pair).

Specifications

Antigen IL-1 beta/IL-1F2
Applications Western Blot, Immunocytochemistry, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. Q63264
Gene Alias catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Gene Symbols IL1B
Host Species Goat
Immunogen E. coli-derived recombinant rat IL-1 beta/IL-1F2 (R&D Systems, Catalog # 501-RL) Val117-Ser268 Accession # Q63264
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3553
Test Specificity Detects rat IL-1 beta/IL-1F2 in ELISAs and Western blots. In sandwich immunoassays, less than 0.8% cross-reactivity with recombinant mouse (rm) IL-1 beta is observed and less than 0.2% cross-reactivity with recombinant human (rh) IL-1 alpha , recombinant rat IL-1 alpha , recombinant porcine (rp) IL-1 alpha , rhIL-1 beta , rpIL-1 beta , rhIL-1 RA, rmIL-1 RA, rpIL-1 RA, rhIL-1 RII, rmIL-1 RI Fc Chimera, rhIL-1 Rrp2 Fc Chimera, and rhIL-1 RAcP Fc Chimera is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.