Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Rat IL-1 RII ELISA Kit (Colorimetric)
SDP

Catalog No. NB231775 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB231775 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB231775 Supplier Novus Biologicals Supplier No. NBP280384

Specifications

Assay Range 15.63-1000 pg/mL
Assay Sensitivity 4.7 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum
Specificity Recognizes both recombinant and natural Rat IL-1 RII.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 7850
Gene Symbol IL1R2
Quantity 1 Kit
Research Discipline Cancer, Phospho Specific, Signal Transduction
Storage Requirements Storage is content dependent.
Synonym beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.