Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IL-17/IL-17A Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
$474.50
Specifications
Antigen | IL-17/IL-17A |
---|---|
Clone | 905515 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB84101100
![]() |
R&D Systems
MAB84101100 |
100 μg |
Each of 1 for $474.50
|
|
|||||
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Rat samples. It is validated for ELISASpecifications
IL-17/IL-17A | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
Chinese hamster ovary cell line CHO-derived recombinant rat IL-17A + rat IL-17F, (Met1-Ser150,I46L) and (Met1-161), Accession # Q61453 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
905515 | |
ELISA | |
Unconjugated | |
Mouse | |
Rat | |
Q61453 | |
3605 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title