Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat IL-17A/F Heterodimer Protein

Catalog No. 9340IL025CF
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat IL-17A/F Heterodimer Protein is derived from CHO. The Recombinant Rat IL-17A/F Heterodimer Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

Specifications

Accession Number Q61453 (IL-17A), Q5BJ95 (IL-17F)
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Molecular Weight (g/mol) 15 kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived rat IL-17A/F Heterodimer protein, (N-terminus)Rat IL-17A (Ala18-Ser150 (Ile46Leu)) Accession &Num; Q61453 Rat IL-17F (Arg29-Ala161) Accession &Num; Q5BJ95 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Disulfide-linked heterodimer
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Biological Activity 3 to 18ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.