Learn More
Description
This kit is designed for the development of solid phase sandwich ELISAs to measure natural and recombinant rat IL-18.
- Assay Type: Solid Phase Sandwich ELISA
- Assay Range: 62.5–4000 pg/mL
- Sample Type: Cell culture supernates, serum, and plasma
- Reactivity: Rat
- Assay Format: 96-well strip plate
- Detection Method: Colorimetric ELISA at 450 nm (TMB)
- Conjugate: Biotin
- Label: HRP
- Cross-reactivity: 16.6% with mouse IL-18
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA |
| Quantity | 1 Kit (for 5 Plates) |
| Species | Rat |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3606 |
| Product Line | R&D Systems |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Research Area | Cytokines |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
