Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IL-18/IL-1F4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB521SP
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Rat samples. It is validated for Western Blot.Specifications
IL-18/IL-1F4 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3606 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot | |
69604 | |
Western Blot 1 ug/mL | |
P97636.1 | |
IL18 | |
E. coli-derived recombinant rat IL-18/IL-1F4 His37-Ser194 Accession # P97636.1 | |
25 μg | |
Primary | |
Detects rat IL-18/IL-1F4 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant mouse IL-18 and no cross-reactivity with recombinant human IL-18, recombinant porcine IL-18, recombinant rat (rr) IL-1 alpha , or rrIL-1 beta . | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction