Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IL-18/IL-1F4 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF521
This item is not returnable.
View return policy
Description
IL-18/IL-1F4 Polyclonal specifically detects IL-18/IL-1F4 in Rat samples. It is validated for Western Blot, Neutralization.Specifications
IL-18/IL-1F4 | |
Polyclonal | |
Unconjugated | |
P97636 | |
IL18 | |
E. coli-derived recombinant rat IL-18/IL-1F4 His37-Ser194 Accession # P97636 | |
100 μg | |
Primary | |
Detects rat IL-18/IL-1F4 in direct ELISAs and Western blots. In Western blots, approximately 35% cross-reactivity with recombinant mouse IL-18 is observed and approximately 10% cross-reactivity with recombinant human IL-18 is observed. | |
Rat | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 1-3 ug/mL | |
Iboctadekin, IFN-gamma-inducing factor, IGIF, IGIFIL-1 gamma, IL18, IL-18MGC12320, IL-1F4, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
Goat | |
Affinity Purified | |
RUO | |
3606 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction