Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IL-1Rrp2/IL-1R6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF573
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF573 100 μg
AF573SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF573 Supplier R&D Systems Supplier No. AF573
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IL-1Rrp2/IL-1R6 Polyclonal specifically detects IL-1Rrp2/IL-1R6 in Rat samples. It is validated for Western Blot.

Specifications

Antigen IL-1Rrp2/IL-1R6
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q62929
Gene Alias IL-1 R6, IL-1 Rrp2, IL1RL2, IL-1RL2, IL1Rrp2, IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2
Gene Symbols Il1rl2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat IL-1 Rrp2/IL-1 R6 Gly22-Tyr340 Accession # Q62929
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8808
Test Specificity Detects rat IL-1 Rrp2/IL-1 R6 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) IL-1 R6 is observed and less than 5% cross-reactivity with rhIL-18 R beta , rhIL-1 RI, and rhIL-1 RII is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.