Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IL-1Rrp2/IL-1R6 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF573
This item is not returnable.
View return policy
Description
IL-1Rrp2/IL-1R6 Polyclonal specifically detects IL-1Rrp2/IL-1R6 in Rat samples. It is validated for Western Blot.Specifications
IL-1Rrp2/IL-1R6 | |
Polyclonal | |
Biotin | |
Q62929 | |
Il1rl2 | |
Mouse myeloma cell line NS0-derived recombinant rat IL-1 Rrp2/IL-1 R6 Gly22-Tyr340 Accession # Q62929 | |
50 μg | |
Primary | |
Detects rat IL-1 Rrp2/IL-1 R6 in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant mouse (rm) IL-18 R is observed, 10% cross-reactivity with recombinant human (rh) IL-1 Rrp2 is observed, and less than 5% cross-reactivity with rmIL-1 RI and rmIL-1 RII is observed. | |
Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
IL-1 R6, IL-1 Rrp2, IL1RL2, IL-1RL2, IL1Rrp2, IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2 | |
Goat | |
Affinity Purified | |
RUO | |
8808 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction