Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IL-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2524SP
This item is not returnable.
View return policy
Description
IL-3 Polyclonal specifically detects IL-3 in Rat samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.Specifications
IL-3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor | |
Goat | |
Affinity Purified | |
RUO | |
3562 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.75-3 ug/mL, Immunocytochemistry 5-15 ug/mL | |
P97688 | |
IL3 | |
E. coli-derived recombinant rat IL-3 Ile27-Cys169 Accession # P97688 | |
25 μg | |
Primary | |
Detects rat IL-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant mouse IL-3 and less than 1% cross-reactivity with recombinant human IL-3 is observed. | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction