Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IL-5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB545SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB545SP 25 μg
MAB545 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB545SP Supplier R&D Systems Supplier No. MAB545SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-5 Monoclonal specifically detects IL-5 in Rat samples. It is validated for Western Blot.

Specifications

Antigen IL-5
Applications Western Blot
Classification Monoclonal
Clone 119905
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q08125
Gene Alias BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I
Gene Symbols IL5
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat IL-5 Met20-Val132 Accession # Q08125
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3567
Test Specificity Detects rat IL-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-5 and recombinant mouse IL-5 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.