Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IL-7R alpha/CD127 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | IL-7R alpha/CD127 |
---|---|
Clone | 717519 |
Dilution | Flow Cytometry 0.25μg/106 cells, CyTOF-ready |
Applications | CyTOF, Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8484SP
![]() |
R&D Systems
MAB8484SP |
25 μg |
Each for $200.50
|
|
|||||
MAB8484100
![]() |
R&D Systems
MAB8484100 |
100 μg |
Each for $502.50
|
|
|||||
Description
IL-7R alpha/CD127 Monoclonal specifically detects IL-7R alpha/CD127 in Rat samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
IL-7R alpha/CD127 | |
Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
Monoclonal | |
Supernatant | |
Rat | |
NP_001099888 | |
3575 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
717519 | |
CyTOF, Flow Cytometry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha | |
Mouse myeloma cell line NS0-derived recombinant rat IL-7 R alpha/CD127 Glu21-Asp239 Accession # NP_001099888 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects rat IL-7 R alpha/CD127in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title