Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IL-7R alpha/CD127 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5607P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5607P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5607P Supplier R&D Systems Supplier No. FAB5607P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

IL-7R alpha/CD127 Polyclonal specifically detects IL-7R alpha/CD127 in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-7R alpha/CD127
Applications Flow Cytometry
Classification Polyclonal
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Accession No. NP_001099888
Gene Alias CD127, CD127 antigen, CD127ILRA, CDW127, IL-7 R alpha, IL-7 receptor subunit alpha, IL7R, IL7R alpha, IL-7R subunit alpha, IL7RA, IL-7RA, IL-7R-alpha, interleukin 7 receptor, interleukin 7 receptor alpha chain, interleukin 7 receptor isoform H5-6, interleukin-7 receptor subunit alpha
Gene Symbols IL7R
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant rat IL-7 R alpha/CD127 Glu21-Asp239 Accession # NP_001099888
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3575
Test Specificity Detects rat IL-7 R alpha/CD127 in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant mouse IL-7 R alpha and less than 10% cross-reactivity with recombinant human IL-7 R alpha is observed.
Target Species Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.