Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IL-9R Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $468.00
Specifications
Antigen | IL-9R |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
IL-9R Polyclonal specifically detects IL-9R in Rat samples. It is validated for Western Blot.Specifications
IL-9R | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Rat | |
CD129, CD129 antigen, IL-9 R, IL-9 receptor, IL9R, IL-9R, interleukin 9 receptor, interleukin-9 receptor | |
IL9R | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
EDM04022 | |
3581 | |
Mouse myeloma cell line NS0-derived recombinant rat IL-9 R Val38-Ala270 Accession # EDM04022 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects rat IL-9 R in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant mouse IL-9 R is observed and approximately 10% cross-reactivity with recombinant human IL-9 R is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title