Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat Jagged 1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF599
This item is not returnable.
View return policy
Description
Jagged 1 Polyclonal specifically detects Jagged 1 in Rat samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
Jagged 1 | |
Polyclonal | |
Biotin | |
Q63722 | |
JAG1 | |
Mouse myeloma cell line NS0-derived recombinant rat Jagged 1 Ser32-Asp1068 (Gly57-Arg59 del, Asp63Thr, Arg64Leu, and Val-Arg-Pro-Tyr ins before Lys69) Accession # Q63722 | |
50 μg | |
Primary | |
Detects rat Jagged 1 in ELISAs and Western blots. In sandwich ELISAs, approximately 14% cross-reactivity with recombinant human (rh) Jagged 1 is observed, and less than 0.3% cross-reactivity with rhJagged 2, recombinant mouse Jadded 2, recombinant rat (rr) Notch, and rrNotch 2 is observed. | |
Rat | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
AGS, AHDMGC104644, Alagille syndrome, AWS, CD339, CD339 antigen, HJ1, JAG1, jagged 1, Jagged1, JAGL1, protein jagged-1 | |
Goat | |
Affinity Purified | |
RUO | |
182 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction