Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat L-Selectin/CD62L Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1534SP
This item is not returnable.
View return policy
Description
L-Selectin/CD62L Polyclonal specifically detects L-Selectin/CD62L in Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
L-Selectin/CD62L | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM1, LAM-1, LAM1LECAM1, LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHR, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L, SELL, TQ1 | |
Goat | |
Affinity Purified | |
RUO | |
6402 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
P30836 | |
SELL | |
Mouse myeloma cell line NS0-derived recombinant rat L-Selectin/CD62L Trp39-Asn332 Accession # P30836 | |
25 μg | |
Primary | |
Detects rat L-Selectin/CD62L in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse L-Selectin and 5% cross-reactivity with recombinant human L-Selectin is observed. | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction