Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat LILRC2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | LILRC2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
LILRC2 Polyclonal specifically detects LILRC2 in Rat samples. It is validated for Western Blot.Specifications
LILRC2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Rat | |
10859 | |
Mouse myeloma cell line NS0-derived recombinant rat LILRC2 Gly24-Asn230 Accession # AAY44812 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects rat LILRC2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant rat LILRC1 is observed. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
AAY44812 | |
Lilrc2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title