Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat LIX Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF543
This item is not returnable.
View return policy
Description
LIX Polyclonal specifically detects LIX in Rat samples. It is validated for Western Blot, Neutralization.Specifications
LIX | |
Polyclonal | |
Unconjugated | |
P97885.1 | |
Goat | |
Affinity Purified | |
RUO | |
6374 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.3-1.5 ug/mL | |
Cxcl6 | |
E. coli-derived recombinant rat LIX Ala38-Gln130 Accession # P97885.1 | |
100 μg | |
Primary | |
Detects rat CXCL5/LIX in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse GCP-2 is observed and less than 5% cross-reactivity with recombinant human GCP-2 is observed and no cross reactivity with _____________ is observed. | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction