Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat M-CSF Antibody, R&D Systems™
Mouse Monoclonal Antibody
Specifications
Antigen | M-CSF |
---|---|
Clone | 427406 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
M-CSF Monoclonal specifically detects M-CSF in Rat samples. It is validated for Western Blot.Specifications
M-CSF | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Rat | |
Q8JZQ0 | |
1435 | |
E. coli-derived recombinant rat M-CSF Glu33-Glu261 Accession # Q8JZQ0 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat M-CSF in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse M-CSF and recombinant human M-CSF is observed. |
427406 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930 | |
CSF1 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title