Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat MAG/Siglec-4a Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB538SP
This item is not returnable.
View return policy
Description
MAG/Siglec-4a Monoclonal specifically detects MAG/Siglec-4a in Rat samples. It is validated for Western Blot, Neutralization.Specifications
MAG/Siglec-4a | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
GMAsialic acid binding Ig-like lectin 4A, myelin associated glycoprotein, myelin-associated glycoprotein, sialic acid-binding immunoglobulin-like lectin 4A, Siglec4a, SIGLEC-4A, S-MAG | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4099 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Neutralization | |
118423 | |
Western Blot 1 ug/mL, Neutralization | |
P07722 | |
MAG | |
Mouse myeloma cell line NS0-derived recombinant rat MAG/Siglec-4a Gly20-Pro516 Accession # P07722 | |
25 μg | |
Primary | |
Detects rat MAG/Siglec-4a in direct ELISAs and Western blots. | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction