Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat MCAM/CD146 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF3250
This item is not returnable.
View return policy
Description
MCAM/CD146 Polyclonal specifically detects MCAM/CD146 in Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
MCAM/CD146 | |
Polyclonal | |
Biotin | |
Q9EPF2 | |
MCAM | |
Mouse myeloma cell line NS0-derived recombinant rat MCAM/CD146 Arg19-Lys560 Accession # Q9EPF2 | |
50 μg | |
Primary | |
Detects rat MCAM/CD146 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human MCAM is observed. | |
Rat | |
IgG |
Western Blot, Flow Cytometry | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, L-Gicerin, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18, MUC18Gicerin | |
Goat | |
Affinity Purified | |
RUO | |
4162 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction