Learn More
Rat MCAM/CD146 PE-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | MCAM/CD146 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 404722 |
| Conjugate | R-PE |
| Dilution | Flow Cytometry 0.25-1 μg/106 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | Q9EPF2 |
| Gene Alias | CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, L-Gicerin, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18, MUC18Gicerin |
| Gene Symbols | MCAM |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.