Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat mGluR2/3 Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Supplier: R&D Systems PPS078
This item is not returnable.
View return policy
Description
mGluR2/3 Polyclonal specifically detects mGluR2/3 in Rat samples. It is validated for Western Blot.Specifications
mGluR2/3 | |
Polyclonal | |
Western Blot 1:1000 dilution | |
GRM2 | |
Peptide from the C-terminus region of rat mGluR2 and mGluR3 | |
100 μL | |
Primary | |
Rat ∼110 kDa mGluR2 and mGluR3 monomers and ∼220 kDa homodimers | |
For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year. |
Western Blot | |
Unconjugated | |
GLUR2, glutamate metabotropic receptor 2, glutamate receptor, metabotropic 2, GPRC1B, GPRC1Bmetabotropic glutamate receptor 2, GPRC1C, GRM2, GRM3, mGlu2, mGluR2, MGLUR2glutamate receptor homolog, mGluR7 | |
Rabbit | |
Affinity Purified | |
RUO | |
2912 | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction