Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat MIS/AMH Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1446
This item is not returnable.
View return policy
Description
MIS/AMH Monoclonal specifically detects MIS/AMH in Rat samples. It is validated for Western Blot.Specifications
MIS/AMH | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P49000 | |
AMH | |
E. coli-derived recombinant rat MIS aa 447-553 Accession # P49000 | |
500 μg | |
Primary | |
Detects rat MIS in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 100% cross-reactivity with recombinant mouse (rm)MIS. | |
Rat | |
Purified |
Western Blot | |
187406 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AMH, MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
268 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction