Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat MMP-8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
| Antigen | MMP-8 |
|---|---|
| Clone | 400102 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
Description
MMP-8 Monoclonal specifically detects MMP-8 in Rat samples. It is validated for Western Blot.Specifications
| MMP-8 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat | |
| O88766 | |
| 4317 | |
| Mouse myeloma cell line NS0-derived recombinant rat MMP-8 Leu21-Pro466 Accession # O88766 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat MMP-8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse MMP-8 and 50% cross-reactivity with recombinant human MMP-8 is observed. |
| 400102 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLG1HNC, Collagenase 2, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL | |
| MMP8 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title