Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Neuropilin-2 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB567RT100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB567RT100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB567RT100 Supplier R&D Systems Supplier No. FAB567RT100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neuropilin-2 Monoclonal specifically detects Neuropilin-2 in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Neuropilin-2
Applications Flow Cytometry
Classification Monoclonal
Clone 96009R
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O35276
Gene Alias MGC126574, neuropilin 2, neuropilin-2, Neuropilin2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17)
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-2, Gln23-Asp857 (Val809-Asp825 del), Accession # O35276
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8828
Target Species Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.