Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat Neuropilin-2 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF567
This item is not returnable.
View return policy
Description
Neuropilin-2 Polyclonal specifically detects Neuropilin-2 in Rat samples. It is validated for Western Blot.Specifications
Neuropilin-2 | |
Polyclonal | |
Biotin | |
O35276 | |
NRP2 | |
Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-2 Gln23-Asp857 (Val809-Asp825 del) Accession # O35276 | |
50 μg | |
Primary | |
Detects rat Neuropilin-2 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant rat Neuropilin-1 is observed. | |
Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
MGC126574, neuropilin 2, neuropilin-2, Neuropilin2, neuropilin-2a(22), neuropilin-2b(0), NP2, NPN 2, NPN2, PRO2714, receptor for VEGF165 and semaphorins class3, Vascular endothelial cell growth factor 165 receptor 2, vascular endothelial growth factor-165 receptor 2, VEGF1265R2, VEGF165R2neuropilin-2a(17) | |
Goat | |
Affinity Purified | |
RUO | |
8828 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction