Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat μ Opioid R/OPRM1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB86291SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB86291SP 25 μg
MAB86291 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB86291SP Supplier R&D Systems Supplier No. MAB86291SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

mu Opioid R/OPRM1 Monoclonal specifically detects mu Opioid R/OPRM1 in Rat samples. It is validated for Western Blot.

Specifications

Antigen mu Opioid R/OPRM1
Applications Western Blot
Classification Monoclonal
Clone 1126D
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide
Gene Accession No. P33535
Gene Alias hMOP, KIAA0403, LMOR, MOP, mOpioidR, MOR, MOR1, M-OR-1, MOR-1, MOR1MOR, Mu opiate receptor, Mu opioid receptor, mu-type opioid receptor, opioid receptor, mu 1, OPRM, OPRM1
Gene Symbols OPRM1
Host Species Rabbit
Immunogen Synthetic peptide with a short amino acid sequence from the N-terminus of rat μ Opioid R/OPRM1 Accession # P33535
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4988
Test Specificity Detects rat μ Opioid R/OPRM1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.