Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat Osteoprotegerin/TNFRSF11B ELISA Kit (Colorimetric)
SDP

Catalog No. NB170613 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170613 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB170613 Supplier Novus Biologicals™ Supplier No. NBP318767
Add to Cart
Add to Cart

Colorimetric ELISA Kit

Specifications

Assay Range 15.63-1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
Specificity This kit recognizes Rat Osteoprotegerin/TNFRSF11B in samples. No significant cross-reactivity or interference between Rat Osteoprotegerin/TNFRSF11B and analogues was observed.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 4982
Gene Symbol TNFRSF11B
Quantity 1 Kit
Research Discipline Apoptosis, Biologically Active Proteins, Cell Biology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.