Learn More
Description
Specifications
Specifications
| Assay Range | 0.47 to 30 ng/mL |
| Assay Sensitivity | 0.28 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Other Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Rat Oxidized LDL in samples. No Significant cross-reactivity or interference between Rat Oxidized LDL and analogues was observed. These kits are designed to determine the Oxidized Low Density Lipoprotein level in Rat biological fluid. Oxidized LDL is a general term for LDL particles with oxidatively modified structural components. As a result from free radical attack, both lipid and protein parts of LDL can be oxidized in the vascular wall. |
| For Use With (Application) | ELISA |
| Host Species | Rat |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Oxidized Low Density Lipoprotein |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
