Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat PBEF/Visfatin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6384
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6384 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6384 Supplier R&D Systems Supplier No. AF6384
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PBEF/Visfatin/NAMPT Polyclonal specifically detects PBEF/Visfatin/NAMPT in Rat samples. It is validated for Western Blot.

Specifications

Antigen PBEF/Visfatin/NAMPT
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q80Z29
Gene Alias EC 2.4.2.12, MGC117256, NAmPRTase, NAMPT, nicotinamide phosphoribosyltransferase, NMPRTase, PBEF1, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN
Gene Symbols NAMPT
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21derived recombinant rat PBEF/Visfatin Gln25-His491 Accession # Q80Z29
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10135
Test Specificity Detects rat PBEF/Visfatin in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse PBEF and recombinant human PBEF is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.