Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat PDGF-AA Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1055
This item is not returnable.
View return policy
Description
PDGF-AA Polyclonal specifically detects PDGF-AA in Rat samples. It is validated for Western Blot.Specifications
PDGF-AA | |
Polyclonal | |
Biotin | |
AAB59693 | |
Pdgfa | |
E. coli-derived recombinant rat PDGF-AA (R&D Systems, Catalog # 1055-AA) Ser87-Arg196 Accession # AAB59693 | |
50 μg | |
Primary | |
Detects rat PDGF-AA in Western blots. In this format, approximately 45% cross-reactivity with recombinant rat PDGF-AB is observed, 35% cross-reactivity with recombinant human PDGF-AA is observed, 5% cross-reactivity with recombinant zebrafish PDGF-AA is observed and less than 1% cross-reactivity with recombinant rat PDGF-BB is observed. | |
Rat | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
PDGF1, PDGF-A, PDGFAA | |
Goat | |
Affinity Purified | |
RUO | |
5154 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction