Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Pentraxin 2/SAP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1895
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1895 100 μg
AF1895SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1895 Supplier R&D Systems Supplier No. AF1895
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Pentraxin 2/SAP Polyclonal specifically detects Pentraxin 2/SAP in Rat samples. It is validated for Western Blot.

Specifications

Antigen Pentraxin 2/SAP
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P23680
Gene Alias 9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component
Gene Symbols APCS
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Pentraxin 2/SAP Gln21-Ser228 Accession # P23680
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 325
Test Specificity Detects rat Pentraxin 2/SAP in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human Pentraxin 2 and recombinant mouse Pentraxin 2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.