Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Pentraxin 2/SAP Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB18951
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB18951 500 μg
MAB18951SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB18951 Supplier R&D Systems Supplier No. MAB18951
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Rat samples. It is validated for Blockade of Receptor-ligand Interaction.

Specifications

Antigen Pentraxin 2/SAP
Applications Blockade of Receptor-ligand Interaction
Classification Monoclonal
Clone 273902
Concentration LYOPH
Conjugate Unconjugated
Dilution Blockade of Receptor-ligand Interaction
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P23680
Gene Alias 9.5S alpha-1-glycoprotein, amyloid P component, serum, APCS, MGC88159, PTX2, PTX2serum amyloid P-component, SAP, SAPpentaxin-related, Serum Amyloid P component
Gene Symbols APCS
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Pentraxin 2/SAP Gln21-Ser228 (predicted) Accession # P23680
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 325
Test Specificity Detects rat Pentraxin 2/SAP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Pentraxin 2 or recombinant mouse Pentraxin 2 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.