Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat Perilipin 1 (aa 502-517) Synthetic Peptide

Catalog No. PEP169
Click to view available options
Quantity:
50 μg

Synthetic Peptide

Rat Perilipin A (residues 502-517) Synthetic Peptide, PEP-169, from Invitrogen.

PEP-169 is a 17 amino acid synthetic peptide whose sequence is derived from rat perilipin A protein. The sequence of this peptide is (amino to carboxy terminus): C-E (502 )-P-I-L-G-R-T-Q-Y-S-Q-L-R-K-K-S (517)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and rat perilipin A, catalog # PA1-1051. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 25629
Species Rat
Purification Method purified
Accession Number P43884
Common Name Rat Perilipin 1 (aa 502-517)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Plin1
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence C-E(502 )-P-I-L-G-R-T-Q-Y-S-Q-L-R-K-K-S(517)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.