Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Phospho-Tyrosine Hydroxylase (S40) Antibody, R&D Systems™

Catalog No. PPS041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS041 Supplier R&D Systems Supplier No. PPS041
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Prepared from rabbit serum by affinity purification via sequential chromatography on phospho and dephosphopeptide coupled columns.

Tyrosine Hydroxylase Polyclonal specifically detects Tyrosine Hydroxylase in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen Tyrosine Hydroxylase (p ser40)
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:1000 dilution, Immunofluorescence 1:1000 dilution
Gene Alias DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase
Gene Symbols TH
Host Species Rabbit
Immunogen Phosphopeptide corresponding to amino acid residues surrounding the phospho-S40 of Tyrosine Hydroxylase
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7054
Test Specificity This antibody is specific for the 60 kDa TH in Western blots of PC-12 cell lysates. Some higher molecular weight bands may be detected depending upon the brain region being studied, protein loads, and the detection methods used. The antibody has three orders of magnitude selectivity over dephospho-TH.
Target Species Rat
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.