Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat Prolactin ELISA Kit (Colorimetric)
SDP

Catalog No. NB397848 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397848 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397848 Supplier Novus Biologicals™ Supplier No. NBP312122
Add to Cart
Add to Cart

Kit (Colorimetric)

Specifications

Assay Range 3.13 to 200 ng/mL
Assay Sensitivity 1.88 ng/mL
Detection Method Sandwich ELISA
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Supernatant
Specificity This kit recognizes Rat Prolactin in samples. No significant cross-reactivity or interference between Rat Prolactin and analogues was observed.
Target Species Rat
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 5617
Gene Symbol PRL
Quantity 1 Kit
Research Discipline Biologically Active Proteins, Cancer, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym prolactin
Target Rat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.