Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat Prolactin R Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1112SP
This item is not returnable.
View return policy
Description
Prolactin R Polyclonal specifically detects Prolactin R in Rat samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
Prolactin R | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRLR, PRL-R, prolactin receptor, prolactin receptor delta 7/11, ProlactinR, secreted prolactin binding protein | |
Goat | |
Affinity Purified | |
RUO | |
5618 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 25 ug/mL | |
Q58DZ7 | |
PRLR | |
Mouse myeloma cell line NS0-derived recombinant rat Prolactin R Gln20-Asp229 Accession # Q58DZ7 | |
25 μg | |
Primary | |
Detects rat Prolactin R in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human Prolactin R. | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction