Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat Prolactin R Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1112SP
This item is not returnable.
View return policy
Description
Prolactin R Monoclonal specifically detects Prolactin R in Rat samples. It is validated for Agonist Activity.Specifications
Prolactin R | |
218616 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRLR, PRL-R, prolactin receptor, prolactin receptor delta 7/11, ProlactinR, secreted prolactin binding protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Detects rat Prolactin R in direct ELISAs and Western blots. Does not cross-react with recombinant human Prolactin R or recombinant mouse Prolactin R. | |
Rat | |
Purified |
Monoclonal | |
LYOPH | |
Agonist Activity 0.07-0.3 ug/mL | |
P05710 | |
PRLR | |
Mouse myeloma cell line NS0-derived recombinant rat Prolactin R Gln20-Asp229 Accession # P05710 | |
25 μg | |
Agonist Activity | |
5618 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction