Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Prolactin R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1112
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1112 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1112 Supplier R&D Systems Supplier No. AF1112
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Prolactin R Polyclonal specifically detects Prolactin R in Rat samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen Prolactin R
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 25 ug/mL
Gene Accession No. Q58DZ7
Gene Alias delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRLR, PRL-R, prolactin receptor, prolactin receptor delta 7/11, ProlactinR, secreted prolactin binding protein
Gene Symbols PRLR
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat Prolactin R Gln20-Asp229 Accession # Q58DZ7
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5618
Test Specificity Detects rat Prolactin R in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human Prolactin R.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.