Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Reg3A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1745
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1745 500 μg
MAB1745SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1745 Supplier R&D Systems Supplier No. MAB1745
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reg3A Monoclonal specifically detects Reg3A in Rat samples. It is validated for Western Blot.

Specifications

Antigen Reg3A
Applications Western Blot
Classification Monoclonal
Clone 250802
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_742074
Gene Alias hepatocarcinoma-intestine-pancreas, HIPpancreatitis-associated protein, Human proislet peptide, Lithostathine 3, pancreatic beta cell growth factor, Pancreatitis-associated protein 1, PAP homologous protein, PAP1FLJ18565, PAP2, PAPPAP-H, PBCGF, proliferation-inducing protein 34, proliferation-inducing protein 42, Reg III-alpha, REG3, REG3A, REG-3-alpha, regenerating islet-derived 3 alpha, regenerating islet-derived protein 3-alpha, Regenerating islet-derived protein III-alpha, REG-III
Gene Symbols REG3A
Host Species Mouse
Immunogen E. coli-derived recombinant rat Reg3A Glu26-Gln174 Accession # NP_742074
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5068
Test Specificity Detects rat Reg3A in direct ELISAs and Western blots. In these formats, shows 100% cross-reactivity with recombinant mouse (rm) Reg3A and no cross-reactivity with rmReg1, recombinant human (rh) Reg1B, recombinant rat Reg2, or rhReg4.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.