Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Reg3B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1996SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1996SP 25 μg
AF1996 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1996SP Supplier R&D Systems Supplier No. AF1996SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Reg3B Polyclonal specifically detects Reg3B in Rat samples. It is validated for Western Blot.

Specifications

Antigen Reg3B
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P25031
Gene Alias PAP1, Reg3B
Gene Symbols Reg3b
Host Species Goat
Immunogen E. coli-derived recombinant rat Reg3B Glu27-Gly175 Accession # P25031
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18489
Test Specificity Detects rat Reg3B in direct ELISAs and Western blots. In direct ELISAs, approxmately 75% cross-reactivity with recombinant mouse (rm) Reg3B, and approximately 20% cross-reactivity with rmReg3G is observed, and less than 2% cross-reactivity with recombinant human (rh) Reg3A, rmReg1, rmReg2, rhReg4, rhReg1A, rmReg3A, and rmReg3D is observed. In Western blots, approximately 40% cross-reactivity with rmReg3B is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.