Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Reg3B Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF1996
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF1996 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF1996 Supplier R&D Systems Supplier No. BAF1996
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Reg3B Polyclonal specifically detects Reg3B in Rat samples. It is validated for Western Blot.

Specifications

Antigen Reg3B
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P25031
Gene Alias PAP1, Reg3B
Gene Symbols Reg3b
Host Species Goat
Immunogen E. coli-derived recombinant rat Reg3B Glu27-Gly175 Accession # P25031
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18489
Test Specificity Detects rat Reg3B in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant rat Reg3 and recombinant mouse (rm) Reg3A is observed and less than 2% cross-reactivity with rmReg1, rmReg2 and recombinant human Reg4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.