Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Synaptotagmin-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB43641SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB43641SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB43641SP Supplier R&D Systems Supplier No. MAB43641SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Synaptotagmin 1 Monoclonal specifically detects Synaptotagmin 1 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen Synaptotagmin 1
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone ASV30
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias DKFZp781D2042, p65, SVP65, synaptotagmin Isynaptotagmin 1, synaptotagmin-1, Synaptotagmin1, SYT, SYT1, SytI
Gene Symbols SYT1
Host Species Mouse
Immunogen Rat brain synaptic plasma membranes
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6857
Test Specificity Detects rat Synaptotagmin-1 in Western blots. Detects Synaptotagmin-1 from rat, mouse, human, chicken, Xenopus, and fish.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.