Learn More
Specifications
Specifications
| Assay Range | 0.16-10 ng/mL |
| Assay Sensitivity | 0.07 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Serum, Plasma |
| Specificity | This kit recognizes Rat Testosterone in samples. No significant cross-reactivity or interference between Rat Testosterone and analogues was observed. |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Research Discipline | Cellular Signaling, Developmental Biology |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | 17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
